Anti-OCRL Rabbit Polyclonal Antibody

Supplier: Proteintech
17695-1-AP-150UL
10091-530EA 402 USD
10091-530
Anti-OCRL Rabbit Polyclonal Antibody
Antibodies
OCRL is also named as INPP5F, OCRL1 and belongs to the 5-phosphatase gene family and that Lowe syndrome represents an inborn error of inositol phosphate metabolism.The protein product of the gene that when mutated is responsible for Lowe syndrome, or oculocerebrorenal syndrome (OCRL), is an inositol polyphosphate 5-phosphatase.It may function in lysosomal membrane trafficking by regulating the specific pool of phosphatidylinositol 4,5-bisphosphate that is associated with lysosomes.It has 2 isoforms produced by alternative splicing.Defects in OCRL are the cause of Lowe oculocerebrorenal syndrome (OCRL) and Dent disease type 2 (DD2).This antibody is specific to OCRL.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: OCRL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR