Anti-ABL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17693-1-AP-150UL
10082-370EA 402 USD
10082-370
Anti-ABL2 Rabbit Polyclonal Antibody
Antibodies
ABL2(Abelson tyrosine-protein kinase 2) is also named as ABLL, ARG and belongs to the protein kinase superfamily. It is a membrane-anchored nonreceptor tyrosine kinase that is a novel mediator of SEMA3F-induced RhoA inactivation and collapsing activity and it has a role in cytoskeletal rearrangements by its C-terminal F-actin- and microtubule-binding sequences. ABL2 can be polyubiquitinated and the polyubiquitination of ABL2 leads to degradation. This protein has 9 isoforms produced by alternative splicing with the molecular weight from 114 kDa to 128 kDa. The antibody is specific to the others except the 60 kDa isoform of ABL2.

Western Blot: HeLa Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IHC: Human Liver cancer Tissue, 1:20-1:200

Type: Primary
Antigen: ABL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR