Anti-PJA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17687-1-AP-150UL
10092-372EA 402 USD
10092-372
Anti-PJA1 Rabbit Polyclonal Antibody
Antibodies
PJA1(E3 ubiquitin-protein ligase Praja-1), also named as RNF70, regulates the level of PRC2 (polycomb repressive complex 2) by targeting its free subunits for Ub-mediated proteasomal degradation and it can be involved in protein ubiquitination in the brain and is a suitable candidate gene for MRX. The Praja1 protein is unlikely to be a membrane receptor, since it lacks a hydrophobic transmembrane domain. It has 3 isoforms produced by alternative splicing with the molecular weight of 71 kDa, 50 kDa and 65 kDa.

Western Blot: Mouse Brain Tissue, 1:200-1:2000; IP: Mouse Brain Tissue, 1:200-1:1000

Type: Primary
Antigen: PJA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR