Anti-SENP3 Rabbit Polyclonal Antibody

Supplier: Proteintech
17659-1-AP-150UL
10094-300EA 402 USD
10094-300
Anti-SENP3 Rabbit Polyclonal Antibody
Antibodies
SENP3(Sentrin-specific protease 3) is also named as SSP3, SUSP3 and belongs to the peptidase C48 family. It acts as an essential factor for ribosome biogenesis. SENP3 might regulate the sumoylation of other substrates through directly binding or mediating association with NPM1 at the nucleolus and other subcellular compartments.

Western Blot:Y79 Cells, 1:500-1:5000

Type: Primary
Antigen: SENP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR