Anti-LGR6 Rabbit Polyclonal Antibody

Supplier: Proteintech
17658-1-AP-150UL
10089-618EA 402 USD
10089-618
Anti-LGR6 Rabbit Polyclonal Antibody
Antibodies
LGR6 (Leucine-rich repeat-containing G-protein coupled receptor 6) is a seven-transmembrane protein. LGR6 is a high affinity receptor of R-spondins that potentiates the canonical Wnt signaling pathway and acts as a marker of multipotent stem cells in the epidermis. It potentially functions as a tumor suppressor.

Western Blot: Mouse Testis Tissue, 1:200-1:1000; IHC: Human Testis Tissue, 1:20-1:200; FC: HeLa Cells, N/A

Type: Primary
Antigen: LGR6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR