Anti-ENPEP Rabbit Polyclonal Antibody

Supplier: Proteintech
17655-1-AP-150UL
10086-536EA 402 USD
10086-536
Anti-ENPEP Rabbit Polyclonal Antibody
Antibodies
IF: HepG2 Cells, 1:10-1:100; FC: HepG2 Cells, N/A

Type: Primary
Antigen: ENPEP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR