Anti-PAAF1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17650-1-AP-150UL
10091-714EA 402 USD
10091-714
Anti-PAAF1 Rabbit Polyclonal Antibody
Antibodies
PAAF1(Proteasomal ATPase-associated factor 1) is also named as WDR71 and belongs to the WD repeat PAAF1/RPN14 family. It functions as a negative regulator of the proteasome by controling the assembly/disassembly of the proteasome. PAAF1 has 3 isoforms produced by alternative splicing.

Western Blot: Mouse Brain Tissue, 1:1000-1:10000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: PAAF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR