Anti-RORB Rabbit Polyclonal Antibody

Supplier: Proteintech
17635-1-AP-150UL
10093-866EA 402 USD
10093-866
Anti-RORB Rabbit Polyclonal Antibody
Antibodies
Nuclear receptors (NRs) are a large group of ligand-inducible transcription factors that regulate many facets of mammalian Physiology . In addition, NRs that lack identified ligands and are termed orphan receptors. RORB is a transcription factor and belongs to the nuclear receptor family , which share a common modular structure composed of a transactivation domain, a DNA-binding domain, and a ligand-binding. Its function as transcriptional factor is regulated by small lipophilic molecules, such as steroid hormones, vitamin D, retinoic acids, and thyroid hormone .

Western Blot:Y79 Cells, 1:200-1:2000

Type: Primary
Antigen: RORB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR