Anti-CDON Rabbit Polyclonal Antibody

Supplier: Proteintech
17634-1-AP-150UL
10084-542EA 402 USD
10084-542
Anti-CDON Rabbit Polyclonal Antibody
Antibodies
Western Blot:SH-SY5Y Cells, 1:500-1:5000

Type: Primary
Antigen: CDON
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR