Anti-NDUFB2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17614-1-AP-150UL
10090-964EA 402 USD
10090-964
Anti-NDUFB2 Rabbit Polyclonal Antibody
Antibodies
The multisubunit NADH:ubiquinone oxidoreductase (complex I) is the first enzyme complex in the electron transport chain of mitochondria. NDUFB2, also named as CI-AGGG, is a nuclear encoded subunit located in the hydrophobic protein fraction of complex I. It may have a role in the regulation of molecular changes during the global cardiac ischemia/reperfusion injury during cardiopulmonary bypass (CPB). The full length 12 kDa protein has a transit peptide with 33 amino acids.

Western Blot: Human Heart Tissue, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: NDUFB2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR