Anti-MED20 Rabbit Polyclonal Antibody

Supplier: Proteintech
17598-1-AP-150UL
10090-092EA 402 USD
10090-092
Anti-MED20 Rabbit Polyclonal Antibody
Antibodies
MED20(Mediator of RNA polymerase II transcription subunit 20) is also named as TRFP and belongs to the Mediator complex subunit 20 family. The deduced 209-amino acid protein shares 44% amino identity with Drosophila Trfp. TRFP had an apparent molecular mass of 28 kD by SDS-PAGE. It is identified TRFP as a component of an RNA polymerase II-SRB complex. The TRFP-containing complex could substitute for core RNA polymerase II in driving basal transcription from a minimal promoter.

Western Blot:K-562 Cells, 1:500-1:5000

Type: Primary
Antigen: MED20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR