Anti-CDAN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17597-1-AP-150UL
10084-410EA 402 USD
10084-410
Anti-CDAN1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: CDAN1
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR