Anti-EXOC5 Rabbit Polyclonal Antibody

Supplier: Proteintech
17593-1-AP-150UL
10086-704EA 402 USD
10086-704
Anti-EXOC5 Rabbit Polyclonal Antibody
Antibodies
EXOC5 (SEC10), a component of the exocyst complex, is a 77-kDa protein with a broad tissue distribution (PubMed: 9119050). The exocyst complex, composed of eight evolutionarily conserved subunits (SEC3, SEC5, SEC6, SEC8, SEC10, SEC15, EXO70, and EXO84), is essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. The complex is also essential for the biogenesis of epithelial cell surface polarity.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IP: Mouse Brain Tissue, 1:200-1:2000; IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: EXOC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR