Anti-PYK2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17592-1-AP-150UL
10093-116EA 402 USD
10093-116
Anti-PYK2 Rabbit Polyclonal Antibody
Antibodies
Proline-rich tyrosine kinase 2 (Pyk2; also known as CAK, RAFTK and CADTK) is a cytoplasmic tyrosine kinase implicated to play a role in several introcellular signaling pathways. It is expressed in many cells and tissues and migrated as a 130 kDa band in Western Blotting analysis.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Kidney Tissue, 1:20-1:200

Type: Primary
Antigen: PYK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR