Anti-HRASLS5 Rabbit Polyclonal Antibody

Supplier: Proteintech
17581-1-AP-150UL
10088-446EA 402 USD
10088-446
Anti-HRASLS5 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Placenta, 1:20-1:200

Type: Primary
Antigen: HRASLS5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR