Anti-ST7L Rabbit Polyclonal Antibody

Supplier: Proteintech
17567-1-AP-150UL
10094-592EA 402 USD
10094-592
Anti-ST7L Rabbit Polyclonal Antibody
Antibodies
ST7 (suppressor of tumorigenicity 7) gene has been proposed to be a tumor suppressor gene, product of which is a multi-pass membrane protein belonging to the ST7 family. ST7L (suppressor of tumorigenicity 7 protein-like) gene encodes a multi-pass membrane protein also belonging to the ST7 family, 8 isoforms produced by alternative splicing have been described . However, the molecular function of this gene product has not been determined so far.

Western Blot: Mouse Testis Tissue, 1:500-1:5000; IP: Mouse Testis Tissue, 1:500-1:5000

Type: Primary
Antigen: ST7L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR