Anti-MTA2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17554-1-AP-150UL
10090-528EA 402 USD
10090-528
Anti-MTA2 Rabbit Polyclonal Antibody
Antibodies
Metastasis-associated protein MTA2, also named as MTA1L1 or PID, is a 668 amino acid protein, which contains one BAH domain, one ELM2 domain, one GATA-type zinc finger and one SANT domain. MTA2 localizes in the nucleus and is widely expressed in many tissues. MTA2 may be involved in the regulation of gene expression as repressor and activator.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: MTA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR