Anti-PTPN18 Rabbit Polyclonal Antibody

Supplier: Proteintech
17551-1-AP-150UL
10093-058EA 402 USD
10093-058
Anti-PTPN18 Rabbit Polyclonal Antibody
Antibodies
PTPN18(Tyrosine-protein phosphatase non-receptor type 18) is also named as BDP1 and belongs to the protein-tyrosine phosphatase family. The PTPN18 cDNA encodes a deduced 459-amino acid protein with no transmembrane domain and a calculated molecular weight of 50 kD. PTPN18 has a PEST motif and shares the greatest homology with members of the PTP-PEST gene family. Northern blot analysis indicated that PTPN18 is expressed not only in brain, but also in colon and several different tumor-derived cell lines.. It has 2 isoforms produced by alternative splicing.

Western Blot:Jurkat Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200

Type: Primary
Antigen: PTPN18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR