Anti-HDAC8 Rabbit Polyclonal Antibody

Supplier: Proteintech
17548-1-AP-150UL
10087-616EA 402 USD
10087-616
Anti-HDAC8 Rabbit Polyclonal Antibody
Antibodies
Histone deacetylases (HDAC) are a class of enzymes that remove the acetyl groups from the lysine residues leading to the formation of a condensed and transcriptionally silenced chromatin. At least 4 classes of HDAC were identified. As a class I HDAC, HDAC 8 was primarily found in the nucleus. It catalyzes the deacetylation of lysine residues in the histone N-terminal tails and represses transcription in large multiprotein complexes with transcriptional co-repressors. This antibody is a rabbit polyclonal antibody raised against full length HDAC8 of human origin.

Western Blot: HeLa Cells, 1:500-1:5000

Type: Primary
Antigen: HDAC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR