Anti-MIER3 Rabbit Polyclonal Antibody

Supplier: Proteintech
17543-1-AP-150UL
10090-222EA 402 USD
10090-222
Anti-MIER3 Rabbit Polyclonal Antibody
Antibodies
The mesoderm induction early response (MIER) protein family is known as fibroblast growth factor (FGF)-regulated immediate-early protein family. Activated by FGF, MIER proteins may have a pivotal role in FGF-regulated cellular activities, indicating a potential influence in the progression of certain cancers . MIER3 localizes to the nucleus and primarily functions as a transcriptional repressor. Rat mammary carcinoma susceptibility 1b (Mcs1b) is a quantitative trait locus on RN02 that confers decreased susceptibility when Copenhagen (COP)-resistant alleles are introgressed into a Wistar Furth (WF)-susceptible genome. MIER3 is a candidate Mcs1b gene .

Western Blot: Mouse Uterus Tissue, 1:200-1:2000

Type: Primary
Antigen: MIER3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR