Anti-PARP9 Rabbit Polyclonal Antibody

Supplier: Proteintech
17535-1-AP-150UL
10091-812EA 402 USD
10091-812
Anti-PARP9 Rabbit Polyclonal Antibody
Antibodies
Poly(ADP-ribosyl)ation is a post-translational modification of proteins mediated by one of the 17 members of the poly(ADP-ribose) polymerases (PARP). PARP-9 belongs to the subfamily of macroPARPs, associating one to three macro domains to the PARP domain. Overexpression of PARP-9 stimulates cell migration in vitro, suggesting a role for PARP-9 in the promotion of malignant B cell migration and dissemination in high risk DLBCL. PARP-9 is also likely a transcription coactivator, its overexpression in B lymphocytes, stimulated by interferon gamma (IFNγ), inducing the transcription of IFNγ-controlled genes.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human spleen Tissue, 1:20-1:200

Type: Primary
Antigen: PARP9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR