Anti-IFT52 Rabbit Polyclonal Antibody

Supplier: Proteintech
17534-1-AP-150UL
10088-630EA 402 USD
10088-630
Anti-IFT52 Rabbit Polyclonal Antibody
Antibodies
IFT52, also named as C20orf9 and NGD5, is a component of IFT complex B which is composed of IFT88, IFT57, TRAF3IP1, IFT52, IFT27, HSPB11 and IFT20. IFT52 binds directly to the IFT81/74/27/25 complex. And IFT52 could mediate the interaction between the two subcomplexes. The MW of IFT52 is about 50-54kd.

Western Blot: Human Testis Tissue, 1:200-1:2000; IF: Hela Cells, 1:10-1:100; IP: HeLa Cells, 1:200-1:2000

Type: Primary
Antigen: IFT52
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR