Anti-CTDSPL2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17532-1-AP-150UL
10085-304EA 402 USD
10085-304
Anti-CTDSPL2 Rabbit Polyclonal Antibody
Antibodies
CTD small phosphatase-like protein 2 (CTDSPL2) is also named as HSPC058 and HSPC129, belongs to the CTDSPL2 family. It is a putative RNA-polymerase II carboxy-terminal domain (CTD) phosphatase, whose specific CTD phosphatase activity was verified in vitro, and may regulate the dynamic phosphorylation of RNAP II CTD.( PMID: 17487459) It can obviously improve the expression of ε- and γ-globin genes in K562 cells and CD34+ cells derived from UCB.( PMID: 20932329) This antibody is a rabbit polyclonal antibody raised against a region of human CTDSPL2.

Western Blot: MCF7 Cells, 1:500-1:5000; IHC: Human endometrial cancer Tissue, 1:20-1:200

Type: Primary
Antigen: CTDSPL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR