Anti-POLR3A Rabbit Polyclonal Antibody

Supplier: Proteintech
17530-1-AP-150UL
10091-860EA 402 USD
10091-860
Anti-POLR3A Rabbit Polyclonal Antibody
Antibodies
Western Blot: MCF7 Cells, 1:200-1:1000

Type: Primary
Antigen: POLR3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR