Anti-RNASEH2B Rabbit Polyclonal Antibody

Supplier: Proteintech
17527-1-AP-150UL
10093-772EA 402 USD
10093-772
Anti-RNASEH2B Rabbit Polyclonal Antibody
Antibodies
Ribonuclease H2 is the major nuclear enzyme degrading cellular RNA/DNA hybrids in eukaryotes and the sole nuclease known to be able to hydrolyze ribonucleotides misincorporated during genomic replication. RNASEH2B and RNASEH2C are the non-catalytic subunits of RNase H2 complex. Defects in RNASEH2B are a cause of Aicardi-Goutieres syndrome type 2 (AGS2), a genetically heterogeneous disease with clinical features as thrombocytopenia, hepatosplenomegaly and elevated hepatic transaminases along with intermittent fever may erroneously suggest an infective process.

Western Blot: Mouse Thymus Tissue, 1:200-1:2000

Type: Primary
Antigen: RNASEH2B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR