Anti-CK8 Rabbit Polyclonal Antibody

Supplier: Proteintech
17514-1-AP-150UL
10085-634EA 402 USD
10085-634
Anti-CK8 Rabbit Polyclonal Antibody
Antibodies
Keratins are a large family of proteins that form the intermediate filament cytoskeleton of epithelial cells, which are classified into two major sequence types. Type I keratins are a group of acidic intermediate filament proteins, including K9–K23, and the hair keratins Ha1–Ha8. Type II keratins are the basic or neutral courterparts to the acidic type I keratins, including K1–K8, and the hair keratins, Hb1–Hb6. KRT8 is often paired with keratin 18 in vivo. This antibody is specifically against KRT8.

Western Blot: MCF7 Cells, 1:200-1:2000; IHC: Human Liver Tissue, 1:20-1:200

Type: Primary
Antigen: CK8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR