Anti-Filensin Rabbit Polyclonal Antibody

Supplier: Proteintech
17492-1-AP-150UL
10087-030EA 402 USD
10087-030
Anti-Filensin Rabbit Polyclonal Antibody
Antibodies
Western Blot: HepG2 Cells, 1:200-1:2000; IF: Hela Cells, 1:10-1:100

Type: Primary
Antigen: Filensin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR