Anti-MAP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17490-1-AP-150UL
10089-936EA 537.7 USD
10089-936
Anti-MAP2 Rabbit Polyclonal Antibody
Antibodies
MAP2 (microtubule-associated protein 2) is a cytoskeleton protein abundant in brain and has important role in neuronal morphogenesis. Multiple high molecular weight (MW) and low molecular weight (MW) MAP2 isoforms are expressed within axons, dendrites, and cell bodies. The expression of MAP2 is regulated in both a tissue- and developmentally specific manner. The 280 kD MAP2B is present throughout rat brain development, and the slightly larger MAP2A appears first during the end of the second week of postnatal life. MAP2C, composed of several bands of about 70 kD, is present during early brain development, and largely disappears from the mature brain except for the retina, olfactory bulb, and cerebellum. MAP2 antibodies have been widely used to mark the neuron or dendrite formation. This antibody can recognize both high MW and low MW isoforms of MAP2.

Western Blot:SH-SY5Y Cells, 1:500-1:5000; IHC: Human Brain Tissue, 1:20-1:200; IF: Hela Cells, 1:10-1:100; IP: Mouse Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: MAP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR