Anti-CLUAP1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17470-1-AP-150UL
10084-888EA 402 USD
10084-888
Anti-CLUAP1 Rabbit Polyclonal Antibody
Antibodies
CLUAP1 (Clusterin-associated protein 1) was originally identified as a protein that interacts with CLU/clusterin, and whose gene is frequently upregulated in colon cancer . Overexpression of CLUAP1 was also observed in osteosarcoma . CLUAP1 is most abundantly expressed in testis among normal tissues . Studies reveal that CLUAP1 has a role in ciliogenenesis and cilia-dependent development .

Western Blot: Mouse Testis Tissue, 1:200-1:2000; IF: MDCK Cells, 1:20-1:200

Type: Primary
Antigen: CLUAP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR