Anti-FASTKD2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17464-1-AP-150UL
10086-880EA 524.48 USD
10086-880
Anti-FASTKD2 Rabbit Polyclonal Antibody
Antibodies
FASTKD2(FAST kinase domain-containing protein 2) is also named as KIAA0971 and belongs to the FAST kinase family.A potential cleavage site of the 79 kDa FASTKD2 precursor protein by the mitochondrial matrix protease (MMP) is present between M52 and Q53 and predicts the mature imported protein from the FASTKD2 precursor to be composed of 642 amino acid residues for a MW of approximately 73 kDa and the clinical and MRI findings of the FASTKD2-associated disease are rather unusual for mitochondrial disorders.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IF:COS7 Cells, 1:20-1:200

Type: Primary
Antigen: FASTKD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR