Anti-IFT140 Rabbit Polyclonal Antibody

Supplier: Proteintech
17460-1-AP-150UL
10088-626EA 537.7 USD
10088-626
Anti-IFT140 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Testis Tissue, 1:200-1:1000; IF: HTERT-RPE1 Cells, 1:20-1:200

Type: Primary
Antigen: IFT140
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR