Anti-Oligophrenin 1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17445-1-AP-150UL
10091-578EA 402 USD
10091-578
Anti-Oligophrenin 1 Rabbit Polyclonal Antibody
Antibodies
The OPHN1 gene encodes a Rho-GTPase activating protein (RhoGAP), and mutations in OPHN1 are responsible for non-specific X-linked mental retardation (NSMR). OPHN1 is highly expressed in the brain and present both pre- and postsynaptically in neurons.

Western Blot: Mouse Brain Tissue, 1:500-1:5000; IHC: Human Colon Tissue, 1:20-1:200

Type: Primary
Antigen: Oligophrenin 1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR