Anti-EVI2A Rabbit Polyclonal Antibody

Supplier: Proteintech
17415-1-AP-150UL
10081-306EA 402 USD
10081-306
Anti-EVI2A Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000; IHC: Human Brain, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: EVI2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR