Anti-SNRPA1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17368-1-AP-150UL
10094-940EA 524.48 USD
10094-940
Anti-SNRPA1 Rabbit Polyclonal Antibody
Antibodies
Small nuclear ribonucleoprotein polypeptide a-prime(SNRPA1) is part of the U2 snRNP particle, which associated with the branch point of the lariat-shaped intron, in intermediate in the cascade of mRNA precursor splicing event. It may be required for cell division, and helps the A' protein to bind stem loop IV of U2 snRNA. This is a rabbit polyclonal antibody raised against the full length SNRPA1 of human origin.

Western Blot: HeLa Cells, 1:1000-1:10000; IHC: Human lung, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: SNRPA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR