Anti-TIMP2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17353-1-AP-150UL
10096-016EA 524.48 USD
10096-016
Anti-TIMP2 Rabbit Polyclonal Antibody
Antibodies
IHC: Human Placenta Tissue, 1:20-1:200; IF: HepG2 Cells, 1:10-1:100; Western Blot:Transfected HEK-293 Cells, 1:500-1:5000

Type: Primary
Antigen: TIMP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR