Anti-DNAJC11 Rabbit Polyclonal Antibody

Supplier: Proteintech
17331-1-AP-150UL
10085-998EA 402 USD
10085-998
Anti-DNAJC11 Rabbit Polyclonal Antibody
Antibodies
DNAJC11 is a member of the J proteins also known as HSP40 family of co-chaperones. Recently DNAJC11 has been found to interact with mitofilin, a mitochondrial inner membrane protein, indicating that it may be a molecular chaperone during mitochondrial protein import and folding.

Western Blot: HeLa Cells, 1:200-1:2000; IHC: Human Skeletal Muscle Tissue, 1:20-1:200

Type: Primary
Antigen: DNAJC11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR