Anti-Pepsinogen I Rabbit Polyclonal Antibody

Supplier: Proteintech
17330-1-AP-150UL
10092-132EA 402 USD
10092-132
Anti-Pepsinogen I Rabbit Polyclonal Antibody
Antibodies
Pepsinogen I belongs to the peptidase A1 family. It shows particularly broad specificity and although bonds involving phenylalanine and leucine are preferred, many others are also cleaved to some extent. The gene endoces a 388 amino acid protein with a 15 amino acid signal peptide and 46 amino acid propeptide. This antibody can recognize PGA3(pepsinogen 3), PGA4(pepsinogen 4), PGA5(pepsinogen 5).

Western Blot:SGC-7901 Cells, 1:500-1:5000; IHC: Human Stomach cancer Tissue, 1:20-1:200

Type: Primary
Antigen: Pepsinogen I
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR