Anti-GSG1L Rabbit Polyclonal Antibody

Supplier: Proteintech
17328-1-AP-150UL
10087-898EA 402 USD
10087-898
Anti-GSG1L Rabbit Polyclonal Antibody
Antibodies
GSG1L is a component of the inner core of AMPAR complex. It modifies AMPA receptor (AMPAR) gating. GSG1L is mainly present on B lymphocytes. This antibody can recognize all the 4 isoforms of GSG1L.

IHC: Human tonsillitis Tissue, 1:20-1:200; Western Blot:Transfected HEK-293 Cells, 1:200-1:2000

Type: Primary
Antigen: GSG1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR