Anti-CD1A Rabbit Polyclonal Antibody

Supplier: Proteintech
17325-1-AP-150UL
10084-286EA 402 USD
10084-286
Anti-CD1A Rabbit Polyclonal Antibody
Antibodies
CD1A is an antigen-presenting protein that binds self and non-self lipid and glycolipid antigens and presents them to T-cell receptors on natural killer T-cells. During protein synthesis and maturation, CD1 family members bind endogenous lipids that are replaced by lipid or glycolipid antigens when the proteins are internalized and pass through endosomes, before trafficking back to the cell surface. CD1A is a 50kd glycoprotein. It has a HeteroDimer form with MW 65-70kd.

Western Blot:K-562 Cells, 1:200-1:2000

Type: Primary
Antigen: CD1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR