Anti-TRPV5 Rabbit Polyclonal Antibody

Supplier: Proteintech
17322-1-AP-150UL
10095-930EA 402 USD
10095-930
Anti-TRPV5 Rabbit Polyclonal Antibody
Antibodies
IF: HEK-293 Cells, 1:10-1:100

Type: Primary
Antigen: TRPV5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR