Anti-ERLIN1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17311-1-AP-150UL
10086-634EA 402 USD
10086-634
Anti-ERLIN1 Rabbit Polyclonal Antibody
Antibodies
Western Blot:rat pancreas Tissue, 1:500-1:5000; IHC: Human Testis Tissue, 1:20-1:200; IP: Mouse Heart Tissue, 1:500-1:5000

Type: Primary
Antigen: ERLIN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR