Anti-AGA Rabbit Polyclonal Antibody

Supplier: Proteintech
17299-1-AP-150UL
10082-590EA 402 USD
10082-590
Anti-AGA Rabbit Polyclonal Antibody
Antibodies
AGA(Aspartylglucosaminidase) is also named as N(4)-(beta-N-acetylglucosaminyl)-L-asparaginase and glycosylasparaginase.The open reading frame of the readthrough transcript would indicate AGA close to 40 kDa.It can be cleaved into 2 chains:the 24 kDa α- and 17 kDa β-subunits of the wild-type AGA,in addition,an aberrant polypeptide with a mol.wt of 21 kDa is also detected as a product of proteolytic cleavage of the mutant precursor molecule.AGA can exsit as a dimer with the mol.wt between 100kDa and 49 kDa and His124 at an interface between two heterodimers of AGA is crucial for the thermodynamically stable oligomeric structure of AGA.. It can also exsit as a heterotetrameric protein of 88 kDa.

IHC: Human Testis Tissue, 1:20-1:200

Type: Primary
Antigen: AGA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR