Anti-RPS4Y1 Rabbit Polyclonal Antibody

Supplier: Proteintech
17296-1-AP-150UL
10093-974EA 402 USD
10093-974
Anti-RPS4Y1 Rabbit Polyclonal Antibody
Antibodies
RPS4Y1, a Y-linked gene in the human, encodes a subunit of ribosomal protein S4, a component of the 40S subunit and 60S subunit. Ribosomal protein S4 is the only ribosomal protein known to be encoded by more than one gene, namely this gene, RPS4Y2 and the ribosomal protein S4, X-linked (RPS4X). RPS4Y maps to a 90-kb segment that has been implicated in Turner syndrome. As a homolog with RPS4X, the antibody can recognize endogenous level of RPS4Y and RPS4X, at 29-32kDa.

Western Blot:DU 145 Cells, 1:200-1:2000; IF: HepG2 Cells, 1:10-1:100

Type: Primary
Antigen: RPS4Y1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR