Anti-BXDC2 Rabbit Polyclonal Antibody

Supplier: Proteintech
17295-1-AP-150UL
10083-740EA 402 USD
10083-740
Anti-BXDC2 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Liver Tissue, 1:500-1:5000; IHC: Human pancreas, 1:20-1:200; IF: HepG2, 1:10-1:100

Type: Primary
Antigen: BXDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR