Anti-GIMAP7 Rabbit Polyclonal Antibody

Supplier: Proteintech
17293-1-AP-150UL
10087-550EA 402 USD
10087-550
Anti-GIMAP7 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000; IF: HepG2 Cells, 1:10-1:100; IP:Jurkat Cells, 1:200-1:2000

Type: Primary
Antigen: GIMAP7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR