Anti-KIF6 Rabbit Polyclonal Antibody

Supplier: Proteintech
17290-1-AP-150UL
10089-244EA 402 USD
10089-244
Anti-KIF6 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Mouse Testis Tissue, 1:200-1:2000; IHC: Human Liver, 1:20-1:200

Type: Primary
Antigen: KIF6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR