Anti-UBE2DNL Rabbit Polyclonal Antibody

Supplier: Proteintech
17287-1-AP-150UL
10096-628EA 402 USD
10096-628
Anti-UBE2DNL Rabbit Polyclonal Antibody
Antibodies
IHC: Human Testis, 1:20-1:200

Type: Primary
Antigen: UBE2DNL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR