Anti-UBE2L6 Rabbit Polyclonal Antibody

Supplier: Proteintech
17278-1-AP-150UL
10096-648EA 402 USD
10096-648
Anti-UBE2L6 Rabbit Polyclonal Antibody
Antibodies
Western Blot:Jurkat Cells, 1:500-1:5000; IF: Hela, 1:10-1:100

Type: Primary
Antigen: UBE2L6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR