Anti-CALML3 Rabbit Polyclonal Antibody

Supplier: Proteintech
17275-1-AP-150UL
10083-982EA 402 USD
10083-982
Anti-CALML3 Rabbit Polyclonal Antibody
Antibodies
Western Blot:A431 Cells, 1:200-1:1000; IF:A431 Cells, 1:20-1:200

Type: Primary
Antigen: CALML3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR