Anti-GSTA4 Rabbit Polyclonal Antibody

Supplier: Proteintech
17271-1-AP-150UL
10087-920EA 402 USD
10087-920
Anti-GSTA4 Rabbit Polyclonal Antibody
Antibodies
Western Blot: Human Brain Tissue, 1:500-1:5000

Type: Primary
Antigen: GSTA4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR